Learn More
Cayman Chemical ANTIBODY KendomycIn 500ug
SDP
Supplier:  Cayman Chemical 11037500 UG
Encompass_Preferred
A macrolide from Streptomyces which displays potent cytotoxicity against several carcinoma cell lines (GI50 < 100 nM); also acts as an endothelin receptor antagonist and has antiosteoporotic activities
Catalog No. 50-295-1090