Learn More
Cayman Chemical ANTIBODY KI8751 5mg
SDP
Supplier:  Cayman Chemical 180045 MG
Encompass_Preferred
A potent, orally available inhibitor of the kinase activity of VEGFR2 (IC50 = 0.9 nM); suppresses the growth of VEGF-stimulated HUVECs at nanomolar concentrations; shows significant anti-tumor activity against assorted human tumor xenografts in nude mice
Catalog No. 50-298-0113