Learn More
Cayman Chemical ANTIBODY KIN1148 5mg
SDP
Supplier:  Cayman Chemical 389805 MG
Encompass_Preferred
An IRF3 activator and adjuvant; increases the mRNA expression of the genes encoding ISG54 and OASL in PH5CH8 cells and increases CXCL10 production in phorbol 12-myristate 13-acetate-differentiated THP-1 monocytes at 10 µM; immunization with a vaccine in combination with KIN1148 increases blood IgG, neutralizing, and hemagglutinin antibody titers, decreases PFUs in the lungs and lung-draining lymph nodes, and increases the survival percentage of mice infected with a lethal dose of influenza virus when used as an adjuvant
Catalog No. 50-291-8786