Learn More
Cayman Chemical ANTIBODY KJ Pyr 9 25mg
SDP
Supplier:  Cayman Chemical 1911625 MG
Encompass_Preferred
A cell-permeable inhibitor of c-Myc (Kd = 6.5 nM) that interferes with Myc-Max complex formation; also potently blocks cell growth directed by overexpression of v-Myc; effective in vivo
Catalog No. 50-298-2078