Learn More
Cayman Chemical ANTIBODY KPT-276 10mg
SDP
Supplier:  Cayman Chemical 2915810 MG
Encompass_Preferred
An inhibitor of XPO1/CRM1; increases survival and reduces spleen weight and white blood cell count in an MV4-11 AML mouse xenograft model at 150 mg/kg; reduces tumor volume and increases survival in a BT 145 glioblastoma mouse xenograft model at 75 mg/kg
Catalog No. 50-293-9907