Learn More
Cayman Chemical ANTIBODY KT195 25mg
SDP
Supplier:  Cayman Chemical 1481825 MG
Encompass_Preferred
A selective inhibitor of ABHD6 (IC50 = 10 nM) with negligible activity against other serine hydrolases such as DAGLβ; used both as a probe to study ABHD6 and as a negative control for studies of DAGLβ
Catalog No. 50-297-3013