Learn More
Cayman Chemical ANTIBODY L-838 417 5mg
SDP
Supplier:  Cayman Chemical 220075 MG
Encompass_Preferred
A partial agonist of α2-, α3-, and α5 subunit-containing GABAA receptors; potentiates GABA-induced currents in LTK- cells expressing α2β3γ2, α3β3γ2, or α5β3γ2 subunit-containing GABAA receptors at 100 nM; increases the mechanical paw withdrawal threshold in a rat model of spinal nerve ligation-induced static allodynia at 10 mg/kg; decreases social avoidance in a novel environment in adult and adolescent rats at 1 and 2 mg/kg, respectively
Catalog No. 50-298-7404