Learn More
Cayman Chemical ANTIBODY L-CnalIn 25mg
SDP
Supplier:  Cayman Chemical 900235725 MG
Encompass_Preferred
An aminooxy analog of ornithine that irreversibly inhibits aminotransferases, including ornithine aminotransferase (Ki = 2 µM); cytotoxic to a range of organisms
Catalog No. 50-289-3749