Learn More
Cayman Chemical ANTIBODY L-LeucIn 100g
SDP
Supplier:  Cayman Chemical 34342100 G
Encompass_Preferred
An essential amino acid; increases basal insulin secretion and decreases glucose-induced insulin release in INS-1E cells at 1, 5, and 10 mM; decreases triglyceride levels and increases cholesterol accumulation in INS-1E cells at 1, 5, and 10 mM; stimulates skeletal muscle protein synthesis in food-deprived or exercised rats
Catalog No. 50-294-8776