Learn More
Cayman Chemical ANTIBODY L-MImosIn 250mg
SDP
Supplier:  Cayman Chemical 14337250 MG
Encompass_Preferred
A non-protein amino acid that chelates iron and copper; inhibits certain enzymes that contain iron or copper, including arginase (IC50 = 3.7 µM)
Catalog No. 50-295-2804