Learn More
Cayman Chemical ANTIBODY L202 10mg
SDP
Supplier:  Cayman Chemical 3784110 MG
Encompass_Preferred
An ionizable cationic lipid (pKa = 6.04); intramuscular administration of LNPs containing L202 and encapsulating nucleoside-modified mRNA encoding pre-fusion stabilized SARS-CoV-2 spike glycoprotein increase plasma levels of IgG antibodies targeting SARS-CoV-2 spike glycoprotein in mice and cynomolgus monkeys
Catalog No. 50-291-6826