Learn More
Fluidigm Corp ANTIBODY LABELING KIT 146ND
SDP
Supplier:  Fluidigm Corp 201146A
Encompass
This Maxpar kit lets you conjugate IgG antibodies with the 146Nd metal isotope, with each kit containing enough material to conjugate 440 antibodies in 100 g amounts.
Catalog No. NC0260959