Learn More
Biotium Antibody Labeling Kits Antibody and Protein Labeling Kits Mix n Stain Antibody Labeling Kits CF488 1/EA
Supplier:  Biotium 92586
Encompass_Preferred
Mix-n-Stain CF Dye Nanobody Thiol Labeling Kits are optimized for labeling nanobodies with bright and photostable CF dyes in 2 hours with no purification Kits tolerate common stabilizers Green fluorescent CF488A has Ex/Em at 490/515 nm One labeling of 20-50 ug Nanobody Thiol per kit
Catalog No. 50-005-01418