Learn More
Biotium Antibody Labeling Kits Antibody and Protein Labeling Kits Mix n Stain Antibody Labeling Kits CF505 1/EA
Supplier:  Biotium 92549
Encompass_Preferred
Mix-n-Stain STORM CF Dye Antibody Labeling Kits are optimized for labeling 50 ug of antibody with a superior CF Dye for STORM in 30 minutes with no purification The kits produce a low degree of labeling that is optimal for super-resolution imaging by STORM CF505 has Ex/Em at 505/519 nm
Catalog No. 50-005-01562