Learn More
Biotium Antibody Labeling Kits Antibody and Protein Labeling Kits Mix n Stain Antibody Labeling Kits CF725 1/EA
Supplier:  Biotium 92583
Encompass_Preferred
Mix-n-Stain Antibody Labeling Kits allow you to label your antibody in 30 minutes without purification Labeling tolerates common buffers and antibody stabilizers CF725 has Ex/Em at 729/750 nm One labeling reaction of 20-50 ug antibody per kit
Catalog No. 50-005-01494