Learn More
Biotium Antibody Labeling Kits Antibody and Protein Labeling Kits Mix n Stain Antibody Labeling Kits CF740 1/EA
Supplier:  Biotium 92576
Encompass_Preferred
Mix-n-Stain Antibody Labeling Kits allow you to label your antibody in 30 minutes without purification Labeling tolerates common buffers and antibody stabilizers CF740 has Ex/Em at 742/767 nm One labeling reaction of 5-20 ug antibody per kit
Catalog No. 50-005-01563