Learn More
Cayman Chemical ANTIBODY LCS-1 10mg
SDP
Supplier:  Cayman Chemical 3523110 MG
Encompass_Preferred
An inhibitor of SOD1 (IC50 = 1.07 µM); inhibits the proliferation of NCI H358 cells, an effect that can be decreased by overexpression of SOD1 (IC50 = 0.8 and 3.5 µM, respectively)
Catalog No. 50-291-0007