Learn More
Cayman Chemical ANTIBODY LDAO 1g
SDP
Supplier:  Cayman Chemical 256991 G
Encompass_Preferred
A zwitterionic detergent; can be used to solubilize membrane proteins; has been used to solubilize and extract colicin A from the plasma membrane, to inactivate enveloped viruses, and, in combination with 10MAG, to encapsulate proteins and nucleic acids dissolved in low viscosity fluids
Catalog No. 50-293-3016