Learn More
Cayman Chemical ANTIBODY LED209 5mg
SDP
Supplier:  Cayman Chemical 168445 MG
Encompass_Preferred
A potent QseC inhibitor that blocks both norepinephrine- and epinephrine-triggered QseC-dependent virulence gene expression at 5 pM in vitro
Catalog No. 50-296-8103