Learn More
Cayman Chemical ANTIBODY LetermovIr 1mg
SDP
Supplier:  Cayman Chemical 175561 MG
Encompass_Preferred
A small molecule inhibitor of human cytomegalovirus viral replication (EC50 = ~5 nM with a selectivity index >15,000) that targets viral DNA cleavage and packaging
Catalog No. 50-298-0406