Learn More
Cayman Chemical ANTIBODY LH 846 10mg
SDP
Supplier:  Cayman Chemical 1768610 MG
Encompass_Preferred
A selective inhibitor of CK1δ (IC50 = 290 nM); inhibits CK1δ-dependent phosphorylation and degradation of PER1 protein and lengthens the circadian period in U2OS cells
Catalog No. 50-296-4895