Learn More
Cayman Chemical ANTIBODY LH1307 5mg
SDP
Supplier:  Cayman Chemical 287285 MG
Encompass_Preferred
An inhibitor of the PD-1/PD-L1 protein interaction (IC50 = 3 nM in an HTRF assay); increases the activation of Jurkat cells expressing PD-1 in co-culture with U2OS or CHO cells expressing PD-L1 (EC50s = 79 and 763 nM, respectively, in reporter assays)
Catalog No. 50-293-8705