Learn More
Cayman Chemical ANTIBODY LIpId 29 10mg
SDP
Supplier:  Cayman Chemical 3533710 MG
Encompass_Preferred
An ionizable cationic amino lipid (pKa = 6.91); administration of human EPO mRNA in lipid 29-containing lipid nanoparticles increases serum human EPO levels in mice
Catalog No. 50-291-3812