Learn More
Cayman Chemical ANTIBODY LIpId 5 10mg
SDP
Supplier:  Cayman Chemical 3437210 MG
Encompass_Preferred
An ionizable cationic lipid (pKa = 6.56); has been used in the generation of LNPs for the delivery of mRNA in vitro and in vivo; intravenous administration of LNPs containing lipid 5 and encapsulating mRNA encoding hEPO increase serum hEPO levels in cynomolgus monkeys
Catalog No. 50-294-8523