Learn More
Cayman Chemical ANTIBODY LIpId A9 50mg
SDP
Supplier:  Cayman Chemical 3766750 MG
Encompass_Preferred
An ionizable cationic lipid (pKa = 6.27); has been used in the generation of LNPs for the delivery of mRNA and siRNA in vivo; LNPs containing lipid A9 and encapsulating non-stimulatory siRNA increase plasma levels of CCL2, indicating activation of the innate immune response, and decrease body weight in mice
Catalog No. 50-291-5964