Learn More
Cayman Chemical ANTIBODY LIpoxIn A4 50UG
SDP
Supplier:  Cayman Chemical 9041050 UG
Encompass_Preferred
A trihydroxy fatty acid containing a conjugated tetraene produced by the metabolism of (±)15-HETE or 15-HpETE with human leukocytes; promotes leukocyte activation, chemotaxis effects, natural killer cell inhibition, and monocyte migration and adhesion
Catalog No. 50-289-3408