Learn More
Cayman Chemical ANTIBODY LOC1886 10mg
SDP
Supplier:  Cayman Chemical 3802610 MG
Encompass_Preferred
A GPX4 inhibitor (Ki = 102 µM); covalently binds to GPX4U46C and inhibits GPX4-mediated reduction of phospholipid hydroperoxides in HT-1080 cell lysates at 100 µM; increases lipid peroxidation in HT-1080 cells at 100 µM
Catalog No. 50-291-7852