Learn More
Cayman Chemical ANTIBODY LomustIn 100mg
SDP
Supplier:  Cayman Chemical 21237100 MG
Encompass_Preferred
A DNA alkylating agent; forms DNA interstrand crosslinks and strand breaks; reduces tumor volume in a melanoma mouse xenograft model at 4-16 mg/kg
Catalog No. 50-298-1160