Learn More
Cayman Chemical ANTIBODY LuotonIn F 1mg
SDP
Supplier:  Cayman Chemical 142541 MG
Encompass_Preferred
A plant alkaloid that demonstrates cytotoxicity against leukemia P-388 cells (IC50 = 2.3 µM) through a mechanism that involves stabilizing the DNA topoisomerase I-DNA complex
Catalog No. 50-295-4433