Learn More
Cayman Chemical ANTIBODY LuzIndole 50mg
SDP
Supplier:  Cayman Chemical 1599850 MG
Encompass_Preferred
A melatonin receptor antagonist that preferentially targets MT1B over MT1A (Ki values = 10-27 and 158-513 nM, respectively); used both in vitro and in vivo to evaluate the roles of melatonin receptor signaling in diverse pathways
Catalog No. 50-296-8897