Learn More
Cayman Chemical ANTIBODY LXH254 10mg
SDP
Supplier:  Cayman Chemical 2823110 MG
Encompass_Preferred
A pan-RAF inhibitor; selectively inhibits B-RAF and C-RAF over a panel of 456 human kinases; inhibits MAPK signaling in tumor cells expressing mutant B-RAFV600; induces tumor regression in various mouse xenograft models
Catalog No. 50-294-1869