Learn More
Cayman Chemical ANTIBODY LXS-196 5mg
SDP
Supplier:  Cayman Chemical 314555 MG
Encompass_Preferred
A PKC inhibitor (IC50s = 1.9 and 0.4 nM for PKCα and PKCθ, respectively); selective for PKCα and PKCθ over GSK3β (IC50 = 3,100 nM); inhibits proliferation of TMD8 B cell lymphoma and 92.1 uveal melanoma cells (IC50s = 900 and 184 nM, respectively) but not SK-MEL-28 skin melanoma cells (IC50 = >10,000 nM)
Catalog No. 50-299-1724