Learn More
Cayman Chemical ANTIBODY LY315920 50mg
SDP
Supplier:  Cayman Chemical 1826750 MG
Encompass_Preferred
An inhibitor of hGIIA sPLA2 (IC50 = 9 nM), as well as mouse GIIA (mGIIA), hGIIE, mGIIE, hGX, and mGX PLA2s, but poorly inhibits other PLA2 enzymes; inhibits serum sPLA2 activity in transgenic mice expressing human sPLA2 when given either intravenously or orally
Catalog No. 50-297-9743