Learn More
Cayman Chemical ANTIBODY LY404039 5mg
SDP
Supplier:  Cayman Chemical 184085 MG
Encompass_Preferred
A potent agonist of mGluR2 and mGluR3 (Kis = 149 and 62 nM, respectively); demonstrates antipsychotic and anxiolytic efficacy in several animal models
Catalog No. 50-296-6524