Learn More
Cayman Chemical ANTIBODY LY404187 25mg
SDP
Supplier:  Cayman Chemical 2874125 MG
Encompass_Preferred
A benzothiadiazide positive allosteric modulator of AMPA receptors; increases glutamate-induced activation of GluR1i, -2i, -2o, -3i, and -4i subunit-containing AMPA receptors with EC50 values of 5.65, 0.15, 1.44, 1.66, and 0.21 µM, respectively, in a calcium influx assay; selective for these AMPA receptors over GluR6 subunit-containing kainate receptors at 10 µM; increases currents induced by glutamate and AMPA in rat prefrontal cortex pyramidal neurons (EC50s = 1.3 and 1.2 µM, respectively), but not in AMPA-stimulated primary rat embryonic hippocampal or primary cerebellar Purkinje neurons; prevents decreases in the number of dopaminergic neurons in the substantia nigra induced by MPTP and 6-OHDA in mouse and rat, respectively, models of Parkinson’s disease at 0.5 mg/kg per day
Catalog No. 50-293-9119