Learn More
Cayman Chemical ANTIBODY M-110 1mg
SDP
Supplier:  Cayman Chemical 391651 MG
Encompass_Preferred
A Pim-3 kinase inhibitor (IC50 = 0.05 µM); selective for Pim-3 over Pim-1 and Pim-2 (IC50s = 2.5 µM for both); inhibits the growth of DU145 and MIA PaCa-2 cells (IC50s = 0.9 and 3.9 µM, respectively); inhibits IL-6-induced STAT3 phosphorylation in DU145 cells at 10 µM
Catalog No. 50-291-6558