Learn More
Cayman Chemical ANTIBODY M50054 1mg
SDP
Supplier:  Cayman Chemical 208101 MG
Encompass_Preferred
A cell-permeable inhibitor of caspase-3 activation; blocks apoptosis in a variety of cell lines induced by different stimuli, including Fas ligand and etoposide
Catalog No. 50-298-3920