Learn More
Cayman Chemical ANTIBODY MCC-555 10mg
SDP
Supplier:  Cayman Chemical 7073510 MG
Encompass_Preferred
A structural homolog of rosiglitazone that binds PPARγ with about 1/10 the affinity of rosiglitazone yet is a more potent antidiabetic agent in whole animal experiments than rosiglitazone (ED50s = 2.7 mg/kg and 7.1 mg/kg, respectively)
Catalog No. 50-289-2450