Learn More
Cayman Chemical ANTIBODY MDK35833 500mg
SDP
Supplier:  Cayman Chemical 34332500 MG
Encompass_Preferred
An inducer of Oct3/4 expression; induces Oct3/4 expression in reporter assays using HEK or NCCIT cells at 10 µM; induces expression of the genes encoding the pluripotency-associated proteins NANOG, REX1, and KLF4 in primary human fibroblasts at 20 µM
Catalog No. 50-294-8381