Learn More
Cayman Chemical ANTIBODY ME0328 10mg
SDP
Supplier:  Cayman Chemical 1848810 MG
Encompass_Preferred
A selective inhibitor of PARP3 (IC50 = 0.89 µM); cell permeable and elicits PARP3-specific effects at submicromolar concentrations
Catalog No. 50-296-6263