Learn More
Cayman Chemical ANTIBODY MEGA-9 500mg
SDP
Supplier:  Cayman Chemical 25702500 MG
Encompass_Preferred
A nonionic detergent that can be used to solubilize membrane proteins; CMC = 20 mM under no-salt conditions; CMCs = 2-17.1 mM under high and low salt conditions for a variety of salts; has been used to solubilize the melibiose transport carrier from E. coli membranes and reconstitute it into liposomes
Catalog No. 50-293-6134