Learn More
Cayman Chemical ANTIBODY -Menthol 100mg
SDP
Supplier:  Cayman Chemical 23463100 MG
Encompass_Preferred
A monoterpene alcohol with antifungal activity; inhibits the growth of F. verticillioides (MIC = 1.5 mM)
Catalog No. 50-299-5292