Learn More
Cayman Chemical ANTIBODY MF438 10mg
SDP
Supplier:  Cayman Chemical 3334910 MG
Encompass_Preferred
A stearoyl-CoA desaturase inhibitor (IC50 = 2.3 nM for the rat enzyme); reduces spheroid formation in NCI H460 and patient-derived NSCLC cells in a concentration-dependent manner; decreases ALDH1A1 activity in NCI H460-derived spheroids at 1 µM; acts synergistically with cisplatin to induce apoptosis and autophagy in patient-derived NSCLC cells
Catalog No. 50-294-8246