Learn More
Cayman Chemical ANTIBODY mG-11 5mg
SDP
Supplier:  Cayman Chemical 387875 MG
Encompass_Preferred
An antagonist of TLR1/2 (IC50 = 0.9 µM in HEK-Blue cells expressing the human receptors); selective for TLR1/2 over the TLR2/6 heterodimer (IC50 = 7.4 µM in HEK-Blue cells expressing the human receptors); inhibits Pam3CKS4-induced TNF-α and IL-6 cytokine secretion in RAW 264.7 macrophages (IC50s = 3.3 and 0.2 µM, respectively); increases SERT and GSH activity, as well as decreases LDH activity and MDA levels, in M. avium-infected Caco-2 cells at 5 ng/ml
Catalog No. 50-291-7059