Learn More
Cayman Chemical ANTIBODY MItoquInol 5mg
SDP
Supplier:  Cayman Chemical 899505 MG
Encompass_Preferred
A mitochondria-targeted antioxidant; reduces hydrogen peroxide production in succinate-fueled isolated bovine aortic endothelial mitochondria
Catalog No. 50-289-4587