Learn More
Cayman Chemical ANTIBODY MK-0941 1mg
SDP
Supplier:  Cayman Chemical 256421 MG
Encompass_Preferred
A glucokinase activator (EC50s = 0.24 and 0.065 µM at low (2.5 mM) and high (10 mM) concentrations of glucose, respectively); increases insulin secretion from isolated rat islets of Langerhans and glucose uptake by hepatocytes in vitro
Catalog No. 50-293-4291