Learn More
Cayman Chemical ANTIBODY MK-1775 5mg
SDP
Supplier:  Cayman Chemical 212665 MG
Encompass_Preferred
An inhibitor of the checkpoint kinase Wee1 (IC50 = 5.2 nM) that prevents phosphorylation of Cdc2 at tryosine15, which abrogates the G2 DNA damage checkpoint; induces apoptosis in combination with several DNA damaging agents selectively in p53-deficient tumor cell lines
Catalog No. 50-298-5457