Learn More
Cayman Chemical ANTIBODY MK-1903 5mg
SDP
Supplier:  Cayman Chemical 343045 MG
Encompass_Preferred
An HCA2 receptor agonist (EC50 = 12.9 nM in an HTRF assay); selective for HCA2 over a panel of GPCRs and ion channels (IC50s = >10 µM for all); reduces plasma free fatty acid levels in fasted rats from 0.001-100 mg/kg
Catalog No. 50-294-9788