Learn More
Cayman Chemical ANTIBODY MK-7246 5mg
SDP
Supplier:  Cayman Chemical 342085 MG
Encompass_Preferred
A CRTH2/DP2 receptor antagonist (Ki = 2.5 nM); selective for CRTH2/DP2 over DP1, FP, IP, and TP receptors (Kis = 373, >25,100, >23,030, and 3,804 nM, respectively), as well as EP1-4 (Kis = 7,668-23,330 nM); inhibits 13,14-dihydro-15-keto-PGD2-induced inhibition of cAMP formation in HEK293 cells expressing human CRTH2/DP2 (IC50 = 3 nM); inhibits 13,14-dihydro-15-keto-PGD2-induced eosinophil shape change (IC50 = 2.2 nM) and CD11b upregulation on eosinophils and basophils in isolated human whole blood (IC50s = 6.2 and 5.4 nM, respectively); decreases antigen-induced late-phase bronchoconstriction and completely prevents airway hyperresponsiveness in sheep at 1 mg/kg
Catalog No. 50-294-6935