Learn More
Cayman Chemical ANTIBODY ML-098 1mg
SDP
Supplier:  Cayman Chemical 151751 MG
Encompass_Preferred
An activator of the GTP-binding protein Rab7 (EC50 = 77.6 nM) that demonstrates selectivity against the related GTPases cdc42, Ras, Rab-2A, and Rac1 (EC50s = 588.8, 346.7, 158.5, and 794.3 nM, respectively)
Catalog No. 50-297-2892